Learn More
Apexbio Technology LLC Piperine 94-62-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092594
Encompass_Preferred
Piperine (CAS 94-62-2) is an alkaloid primarily found in black pepper (Piper nigrum) that has demonstrated diverse pharmacological activities It modulates several signaling pathways including inhibition of drug-metabolizing enzymes such as CYP3A4 and P-glycoprotein and affects cellular processes through regulation of MAPK and Akt phosphorylation Piperine has been studied for its potential to enhance the bioavailability of various therapeutic agents and for its anti-inflammatory anti-tumor and neuroprotective properties This compound is utilized in biomedical research to investigate drug-drug interactions signal transduction mechanisms and its impact on cellular proliferation and apoptosis
Catalog No. 50-000-92594 Supplier Apexbio Technology LLC Supplier No. N16745.1
Piperine (CAS 94-62-2) is an alkaloid primarily found in black pepper (Piper nigrum) that has demonstrated diverse pharmacological activities It modulates several signaling pathways including inhibition of drug-metabolizing enzymes such as CYP3A4 and P-glycoprotein and affects cellular processes through regulation of MAPK and Akt phosphorylation Piperine has been studied for its potential to enhance the bioavailability of various therapeutic agents and for its anti-inflammatory anti-tumor and neuroprotective properties This compound is utilized in biomedical research to investigate drug-drug interactions signal transduction mechanisms and its impact on cellular proliferation and apoptosis