Learn More
Medchemexpress LLC Piperine | 94-62-2 | 98.99% | 285.34 | 100 G
SDP
Catalog No. 5000428085
Encompass_Preferred
Piperine is an alkaloid isolated from pepper. It can inhibit the activity of P-glycoprotein and CYP3A4, and inhibits HeLa cells with an IC50 of 61.94±0.054 μg/mL.
  • Alkaloid isolated from pepper.
  • Inhibits the activity of P-glycoprotein and CYP3A4.
  • Inhibits HeLa cells with an IC50 of 61.94±0.054 μg/mL.
  • Shown to possess in vitro cytotoxic activity.
  • Immunomodulatory, anti-oxidant, anti-asthmatic, anti-carcinogenic, anti-inflammatory, anti-ulcer, and anti-amoebic properties.
  • Enhances the bioavailabilities of other drugs, including rosuvastatin, puerarin, and docetaxel (DOX) by inhibiting CYP3A and P-glycoprotein activity.
  • Inhibits the phosphorylation of I-κB, p65, p38, ERK, and JNK in a dose-dependent manner, suggesting potential as an anti-inflammatory drug.
Catalog No. 50-004-28085 Supplier Medchemexpress LLC Supplier No. HYN0144100G
Piperine is an alkaloid isolated from pepper. It can inhibit the activity of P-glycoprotein and CYP3A4, and inhibits HeLa cells with an IC50 of 61.94±0.054 μg/mL.
  • Alkaloid isolated from pepper.
  • Inhibits the activity of P-glycoprotein and CYP3A4.
  • Inhibits HeLa cells with an IC50 of 61.94±0.054 μg/mL.
  • Shown to possess in vitro cytotoxic activity.
  • Immunomodulatory, anti-oxidant, anti-asthmatic, anti-carcinogenic, anti-inflammatory, anti-ulcer, and anti-amoebic properties.
  • Enhances the bioavailabilities of other drugs, including rosuvastatin, puerarin, and docetaxel (DOX) by inhibiting CYP3A and P-glycoprotein activity.
  • Inhibits the phosphorylation of I-κB, p65, p38, ERK, and JNK in a dose-dependent manner, suggesting potential as an anti-inflammatory drug.