Learn More
Medchemexpress LLC Piperine | 94-62-2 | 99.0% | 285.34 | 10 MM * 1 ML
SDP
Catalog No. 5000427887
Encompass_Preferred
Piperine is an alkaloid isolated from pepper, which inhibits the activity of P-glycoprotein and CYP3A4. It has demonstrated in vitro cytotoxic activity and immunomodulatory, anti-oxidant, anti-asthmatic, anti-carcinogenic, anti-inflammatory, anti-ulcer, and anti-amoebic properties. It can enhance the bioavailability of other drugs.
  • Inhibits activity of P-glycoprotein and CYP3A4
  • Exhibits in vitro cytotoxic activity
  • Possesses immunomodulatory, anti-oxidant, anti-asthmatic, anti-carcinogenic, anti-inflammatory, anti-ulcer, and anti-amoebic properties
  • Enhances bioavailability of other drugs
  • Inhibits phosphorylation of I-κB, p65, p38, ERK, and JNK
Catalog No. 50-004-27887 Supplier Medchemexpress LLC Supplier No. HYN014410MM/1ML
Piperine is an alkaloid isolated from pepper, which inhibits the activity of P-glycoprotein and CYP3A4. It has demonstrated in vitro cytotoxic activity and immunomodulatory, anti-oxidant, anti-asthmatic, anti-carcinogenic, anti-inflammatory, anti-ulcer, and anti-amoebic properties. It can enhance the bioavailability of other drugs.
  • Inhibits activity of P-glycoprotein and CYP3A4
  • Exhibits in vitro cytotoxic activity
  • Possesses immunomodulatory, anti-oxidant, anti-asthmatic, anti-carcinogenic, anti-inflammatory, anti-ulcer, and anti-amoebic properties
  • Enhances bioavailability of other drugs
  • Inhibits phosphorylation of I-κB, p65, p38, ERK, and JNK