Learn More
Medchemexpress LLC Piperine (E/Z-bioperine) | 7780-20-3 | MFCD00005839 | 98.9% | 285.34 g/mol | C17H19NO3 | 250 MG
SDP
Catalog No. 5000280758
Encompass_Preferred
(E/Z)-Piperine is an alkaloid research reagent isolated from black pepper that modulates drug metabolism and transport. It inhibits P-glycoprotein and CYP3A4 and has reported anti-inflammatory, immunomodulatory, neuroprotective, and anticancer activities. Supplied as a light yellow solid for laboratory research use.
  • High purity: 98.9%.
  • Light yellow solid suitable for research applications.
  • Inhibits P-glycoprotein and CYP3A4, useful for metabolism studies.
  • Reported anti-inflammatory and immunomodulatory effects.
  • Available in laboratory pack sizes including 250 mg.
Catalog No. 50-002-80758 Supplier Medchemexpress LLC Supplier No. HYW230975250MG
(E/Z)-Piperine is an alkaloid research reagent isolated from black pepper that modulates drug metabolism and transport. It inhibits P-glycoprotein and CYP3A4 and has reported anti-inflammatory, immunomodulatory, neuroprotective, and anticancer activities. Supplied as a light yellow solid for laboratory research use.
  • High purity: 98.9%.
  • Light yellow solid suitable for research applications.
  • Inhibits P-glycoprotein and CYP3A4, useful for metabolism studies.
  • Reported anti-inflammatory and immunomodulatory effects.
  • Available in laboratory pack sizes including 250 mg.