Learn More
Cayman Chemical Piperitenone, Purity: ≥95%, CAS: 491-09-8, 10 mg
SDP
Catalog No. 502936612
Encompass_Preferred
A monoterpene; scavenges DPPH radicals in a cell free assay (IC50 = 22.7 μg/ml); inhibits peroxidation of linoleic acid
Catalog No. 50-293-6612 Supplier Cayman Chemical Supplier No. 2575210 MG
A monoterpene; scavenges DPPH radicals in a cell free assay (IC50 = 22.7 μg/ml); inhibits peroxidation of linoleic acid