Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperitone, ≥98.0% (mixture of enantiomers, predominantly (-)-enantiomer, GC), MilliporeSigma™ Supelco™

Catalog No. 111015762 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5762 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5762 Supplier MilliporeSigma Supelco Supplier No. 7989910MG
Add to Cart
Add to Cart

Piperitone is a monocyclic monoterpenoid, which occurs in eucalyptus oil. It is a good source of menthone and thymol.

TRUSTED_SUSTAINABILITY

Specifications

CAS 89-81-6
Flash Point 83.5°C - Closed Cup
Linear Formula C10H16 O
MDL Number MFCD00045532
Synonym 3-Carvomenthenone; 6-Isopropyl-3-methyl-2-cyclohexen-1-one; p-Menth-1-en-3-one
Type Analytical Standard
Quantity 10 mg
Formula Weight 152.23
Percent Purity ≥98.0% (mixture of enantiomers, predominantly (to) to enantiomer, GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.