Learn More
AdipoGen Piperlongumine (50 mg)
SDP
Supplier:  AdipoGen AGCN20024M050
Encompass_Preferred
Chemical. CAS: 20069-09-4. Formula: C17H19NO5. MW: 317.3. Isolated from Piper longum roots. Cytotoxic against tumor cell lines. Induces necrosis and apoptosis in cancer cells. Shows anti-platelet aggregation activity possibly by inhibition of cyclooxgenase activity and a decrease in thromboxane A2 formation. Shows significant anxiolytic and antidepressant activities. Promotes adipogenesis of 3T3-L1 cells. Induces in vivo and in vitro mutagenicity in eukaryotic models. Selectively kills cancer cells by targeting the stress response to ROS. Shows in vitro schistosomicidal activity.
Catalog No. 50-205-3369