Learn More
Cayman Chemical Piperlonguminine, Purity: ≥98%, CAS: 5950-12-9, 10 mg
SDP
Catalog No. 502894920
Encompass_Preferred
An alkaloid amide from species of the genus Piper, a plant used in traditional medicine that demonstrates antifungal, anticancer, antihyperlipidemic, and anti-inflammatory properties
Catalog No. 50-289-4920 Supplier Cayman Chemical Supplier No. 900115510 MG
An alkaloid amide from species of the genus Piper, a plant used in traditional medicine that demonstrates antifungal, anticancer, antihyperlipidemic, and anti-inflammatory properties