Learn More
Cayman Chemical Piperlonguminine, Purity: ≥98%, CAS: 5950-12-9, 50 mg
SDP
Catalog No. 502895036
Encompass_Preferred
An alkaloid amide from species of the genus Piper, a plant used in traditional medicine that demonstrates antifungal, anticancer, antihyperlipidemic, and anti-inflammatory properties
Catalog No. 50-289-5036 Supplier Cayman Chemical Supplier No. 900115550 MG
An alkaloid amide from species of the genus Piper, a plant used in traditional medicine that demonstrates antifungal, anticancer, antihyperlipidemic, and anti-inflammatory properties