Learn More
Medchemexpress LLC Piperonyl alcohol | 495-76-1 | MFCD00005836 | 99.8% | 152.15 | C8H8O3 | 1 ML
SDP
Catalog No. 5000300232
Encompass_Preferred
Piperonyl alcohol (CAS 495-76-1) is a benzodioxole-derived alcohol used in research and chemical synthesis. It is biocompatible and can act as an initiator for certain synthetic reactions. The compound is available as a ready-to-use 10 mM solution in DMSO and in multiple solid pack sizes to support small-scale screening and larger preparative work.
  • Biocompatible benzodioxole derivative suitable for research applications.
  • Functions as an initiator in chemical synthesis.
  • Available as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • Offered in multiple solid pack sizes for experimental scaling.
  • High reported purity supports reproducible experimental results.
Catalog No. 50-003-00232 Supplier Medchemexpress LLC Supplier No. HYW02810810MM/1ML
Piperonyl alcohol (CAS 495-76-1) is a benzodioxole-derived alcohol used in research and chemical synthesis. It is biocompatible and can act as an initiator for certain synthetic reactions. The compound is available as a ready-to-use 10 mM solution in DMSO and in multiple solid pack sizes to support small-scale screening and larger preparative work.
  • Biocompatible benzodioxole derivative suitable for research applications.
  • Functions as an initiator in chemical synthesis.
  • Available as a ready-to-use 10 mM solution in DMSO for convenient dosing.
  • Offered in multiple solid pack sizes for experimental scaling.
  • High reported purity supports reproducible experimental results.