Learn More
Chem Service Inc Piperonyl butoxide,51-03-6,100MG
SDP
Catalog No. NC9954965
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula C19H30O5, Purity 97.00%, Molecular Weight 338.43, Boiling Point 180 C, Synonyms: 2-(2-butoxyethoxy)ethyl 6-propyl piperonyl ether
Catalog No. NC9954965 Supplier Chem Service Inc Supplier No. N13061100MG
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula C19H30O5, Purity 97.00%, Molecular Weight 338.43, Boiling Point 180 C, Synonyms: 2-(2-butoxyethoxy)ethyl 6-propyl piperonyl ether