Learn More
Selleck Chemical LLC Piperonyl butoxide S4831-25mg
SDP
Catalog No. 503108982
Encompass_Preferred
Piperonyl butoxide (PBO Butacide Ethanol butoxide Pyrenone 606 ENT-14250) is a man-made pesticide synergist working with insect killers to increase their effectiveness Piperonyl butoxide is an inhibitor of cytochrome P450 monooxygenases(P450s)
Catalog No. 50-310-8982 Supplier Selleck Chemical LLC Supplier No. S483125MG
Piperonyl butoxide (PBO Butacide Ethanol butoxide Pyrenone 606 ENT-14250) is a man-made pesticide synergist working with insect killers to increase their effectiveness Piperonyl butoxide is an inhibitor of cytochrome P450 monooxygenases(P450s)