Learn More
Cayman Chemical Piperonyl Butoxide, Purity: ≥95%, CAS: 51-03-6, 50 mg
SDP
Catalog No. 502936963
Encompass_Preferred
An insecticide synergist; inhibits CYP activity in western corn rootworm abdominal microsomes; increases the toxicity of deltamethrin to field-collected, deltamethrin-resistant strains of the bed bug C. lectularius by 5.6- to 176-fold at 50 μg/μl; increases the toxicity of cyhexatin to fourth instar larvae of susceptible strains of the cotton leafworm S. littoralis; not toxic to the freshwater invertebrates H. azteca, C. tentans, and L. variegatus (LC50s = 530, 2,740, and 3,540 μg/L, respectively) and reduces the toxicity of diazinon, chlorpyrifos, and azinphos-methyl
Catalog No. 50-293-6963 Supplier Cayman Chemical Supplier No. 2582050 MG
An insecticide synergist; inhibits CYP activity in western corn rootworm abdominal microsomes; increases the toxicity of deltamethrin to field-collected, deltamethrin-resistant strains of the bed bug C. lectularius by 5.6- to 176-fold at 50 μg/μl; increases the toxicity of cyhexatin to fourth instar larvae of susceptible strains of the cotton leafworm S. littoralis; not toxic to the freshwater invertebrates H. azteca, C. tentans, and L. variegatus (LC50s = 530, 2,740, and 3,540 μg/L, respectively) and reduces the toxicity of diazinon, chlorpyrifos, and azinphos-methyl