Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonylamine 97.0+%, TCI America™
SDP

Catalog No. P105725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P105725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P105725G Supplier TCI America Supplier No. P105725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2620-50-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00005840
InChI Key ZILSBZLQGRBMOR-UHFFFAOYSA-N
Synonym piperonylamine, benzo d 1,3 dioxol-5-ylmethanamine, 3,4-methylenedioxybenzylamine, 1,3-benzodioxol-5-ylmethylamine, 1,3-benzodioxole-5-methanamine, 2h-1,3-benzodioxol-5-ylmethanamine, 3,4-methylenedioxy benzylamine, benzo-1,3-dioxole-5-methylamine, 1-1,3-benzodioxol-5-yl methanamine, 5-aminomethyl-1,3-benzodioxole
PubChem CID 75799
IUPAC Name 1-(2H-1,3-benzodioxol-5-yl)methanamine
SMILES NCC1=CC=C2OCOC2=C1

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 g
UN Number 2735
Formula Weight 151.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Piperonylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.