Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Piperonylbutoxide

Catalog No. 291102100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-110-2100ML 100 mL
29-110-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-110-2100ML Supplier Sigma Aldrich Supplier No. 291102100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Piperonylbutoxide is a safe insecticide for household and field
  • Effect of piperonyl butoxide, a synthetic methylenedioxyphenyl inhibitor of cytochrome(s) p450, on the toxicity of organophosphate insecticides to cladoceran test species (ceriodaphnia dubia, daphnia magna and daphnia pulex) has been investigated

Specifications

CAS 51-03-6
Density 1.059 g/mL (at 25°C (literature))
Boiling Point 155°C (0.3 mmHg)
Molecular Formula C19H30O5
Linear Formula C19H30O5
Refractive Index n20/D 1.498 (literature)
MDL Number MFCD00005842
Quantity 100 mL
Synonym 2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether; 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether
Molecular Weight (g/mol) 338.44
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.