Learn More
Apexbio Technology LLC PIPERONYLIC-ACID-500MG
SDP
Catalog No. 5000568726
Encompass_Preferred
Piperonylic acid (CAS No 94-53-1) is a compound frequently used in biomedical research particularly in the study of enzyme inhibition and metabolic pathways Originating from the oxidation of piperonal it acts as an inhibitor of cytochrome P450 enzymes which play a crucial role in drug metabolism and detoxification processes Its in vitro activity is commonly observed in the nanomolar to low micromolar range demonstrating high potency Piperonylic acid is utilized in cellular models to evaluate metabolic interactions and investigate enzyme-mediated drug interactions Additionally it is employed in animal studies to assess its potential therapeutic effects and toxicological profiles In drug screening it serves as a tool to slow down or alter specific pathways providing insight into drug efficacy and safety profiles Experimental concentrations typically depend on the specific research design allowing flexibility in various investigative protocols
Catalog No. 50-005-68726 Supplier Apexbio Technology LLC Supplier No. C6537500
Piperonylic acid (CAS No 94-53-1) is a compound frequently used in biomedical research particularly in the study of enzyme inhibition and metabolic pathways Originating from the oxidation of piperonal it acts as an inhibitor of cytochrome P450 enzymes which play a crucial role in drug metabolism and detoxification processes Its in vitro activity is commonly observed in the nanomolar to low micromolar range demonstrating high potency Piperonylic acid is utilized in cellular models to evaluate metabolic interactions and investigate enzyme-mediated drug interactions Additionally it is employed in animal studies to assess its potential therapeutic effects and toxicological profiles In drug screening it serves as a tool to slow down or alter specific pathways providing insight into drug efficacy and safety profiles Experimental concentrations typically depend on the specific research design allowing flexibility in various investigative protocols