Learn More
Medchemexpress LLC Piperoxan hydrochloride | 135-87-5 | 99.7% | 269.77 | C14H20ClNO2 | 10 MG
SDP
Catalog No. 5000212220
Encompass_Preferred
Piperoxan hydrochloride is a research chemical described as an α2 adrenoceptor antagonist and an early-generation antihistamine. Supplied as a solid for laboratory use, it is used as a reference compound in receptor pharmacology and biochemical studies.
  • α2 adrenoceptor antagonist activity
  • First-generation antihistamine properties
  • High reported purity (about 99.7%) suitable for research
  • Solid form supplied in small laboratory pack sizes
  • Molecular formula C14H20ClNO2; molecular weight 269.77
Catalog No. 50-002-12220 Supplier Medchemexpress LLC Supplier No. HY10085010MG
Piperoxan hydrochloride is a research chemical described as an α2 adrenoceptor antagonist and an early-generation antihistamine. Supplied as a solid for laboratory use, it is used as a reference compound in receptor pharmacology and biochemical studies.
  • α2 adrenoceptor antagonist activity
  • First-generation antihistamine properties
  • High reported purity (about 99.7%) suitable for research
  • Solid form supplied in small laboratory pack sizes
  • Molecular formula C14H20ClNO2; molecular weight 269.77