Learn More
Medchemexpress LLC Piperoxan hydrochloride | 135-87-5 | 99.7% | 269.77 g·mol^-1 | C14H20ClNO2 | 100 MG
SDP
Catalog No. 5000211280
Encompass_Preferred
Piperoxan hydrochloride is a research chemical described as an α2 adrenoceptor antagonist and a first-generation antihistamine, provided for laboratory pharmacology and biochemical studies. Consult the product datasheet and safety data sheet for handling, storage, and purity specifications.
  • High reported purity (about 99.7%).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Suitable for pharmacology and biochemical research applications.
  • CAS number provided for unambiguous identification.
  • Manufacturer datasheet and SDS available for safety and handling.
Catalog No. 50-002-11280 Supplier Medchemexpress LLC Supplier No. HY100850100MG
Piperoxan hydrochloride is a research chemical described as an α2 adrenoceptor antagonist and a first-generation antihistamine, provided for laboratory pharmacology and biochemical studies. Consult the product datasheet and safety data sheet for handling, storage, and purity specifications.
  • High reported purity (about 99.7%).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Suitable for pharmacology and biochemical research applications.
  • CAS number provided for unambiguous identification.
  • Manufacturer datasheet and SDS available for safety and handling.