Learn More
Medchemexpress LLC Piperoxan hydrochloride | 135-87-5 | MFCD00079224 | ≥98% | 269.77 g/mol | C14H20ClNO2 | 1 ML
SDP
Catalog No. 5000211281
Encompass_Preferred
Piperoxan hydrochloride is an α2 adrenoceptor antagonist and first-generation antihistamine supplied as a 10 mM solution in DMSO (1 mL) for research use. It is identified by CAS 135-87-5 and has a molecular weight of 269.77 g/mol. The ready-to-use format is intended for in vitro receptor pharmacology and small-scale assay applications.
  • Ready-to-use 10 mM solution in DMSO.
  • Suitable for adrenergic receptor and antihistamine research.
  • Supplied at 1 mL volume for small-scale assays.
  • Molecular weight 269.77 g/mol; CAS 135-87-5.
  • Reported high purity (≥98%) in supplier documentation.
Catalog No. 50-002-11281 Supplier Medchemexpress LLC Supplier No. HY10085010MM 1ML
Piperoxan hydrochloride is an α2 adrenoceptor antagonist and first-generation antihistamine supplied as a 10 mM solution in DMSO (1 mL) for research use. It is identified by CAS 135-87-5 and has a molecular weight of 269.77 g/mol. The ready-to-use format is intended for in vitro receptor pharmacology and small-scale assay applications.
  • Ready-to-use 10 mM solution in DMSO.
  • Suitable for adrenergic receptor and antihistamine research.
  • Supplied at 1 mL volume for small-scale assays.
  • Molecular weight 269.77 g/mol; CAS 135-87-5.
  • Reported high purity (≥98%) in supplier documentation.