Learn More
Medchemexpress LLC Piperoxan hydrochloride | 135-87-5 | MFCD00079224 | 99.7% | 269.77 | C14H20ClNO2 | 5 MG
SDP
Catalog No. 5000211492
Encompass_Preferred
Piperoxan hydrochloride is a small-molecule research reagent described as an α2 adrenoceptor antagonist and a first-generation antihistamine. Supplied as the hydrochloride salt (CAS 135-87-5), it is used in pharmacology and receptor-binding studies to probe adrenergic and histaminergic mechanisms.
  • High purity research-grade small molecule (molecular weight 269.77).
  • Acts as an α2 adrenoceptor antagonist for receptor pharmacology studies.
  • Exhibits first-generation antihistamine properties relevant to histamine assays.
  • Available in milligram-scale quantities suitable for in vitro research.
  • Provided with SDS and COA documentation for handling and quality assurance.
Catalog No. 50-002-11492 Supplier Medchemexpress LLC Supplier No. HY1008505MG
Piperoxan hydrochloride is a small-molecule research reagent described as an α2 adrenoceptor antagonist and a first-generation antihistamine. Supplied as the hydrochloride salt (CAS 135-87-5), it is used in pharmacology and receptor-binding studies to probe adrenergic and histaminergic mechanisms.
  • High purity research-grade small molecule (molecular weight 269.77).
  • Acts as an α2 adrenoceptor antagonist for receptor pharmacology studies.
  • Exhibits first-generation antihistamine properties relevant to histamine assays.
  • Available in milligram-scale quantities suitable for in vitro research.
  • Provided with SDS and COA documentation for handling and quality assurance.