Learn More
Medchemexpress LLC Piperoxan hydrochloride | 135-87-5 | MFCD00079224 | 99.7% | 269.77 g/mol | C14H20ClNO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10085050MG
Encompass_Preferred
Piperoxan hydrochloride is a small-molecule research reagent that functions as an α2 adrenoceptor antagonist and an early-generation antihistamine. Supplied as the hydrochloride salt in high purity, it is used in receptor pharmacology, GPCR studies, and biochemical assays, and is commonly provided as small solid quantities or prepared DMSO stock solutions.
  • α2 adrenoceptor antagonist activity suitable for receptor pharmacology studies.
  • First-generation antihistamine properties for comparative pharmacology.
  • High purity (manufacturer-reported ~99.7%) appropriate for biochemical assays.
  • Available as small solid quantities and as prepared DMSO solutions for convenience.
  • Molecular formula C14H20ClNO2 and molecular weight 269.77 g/mol.
  • Safety and quality documents available, including SDS and COA.
Catalog No. 50-002-12290