Learn More
Cayman Chemical PIperoxnhydrochlorIde 10mg
SDP
Catalog No. 502940642
Encompass_Preferred
An α2-AR antagonist (Kis = 5.4, 2, and 1.3 nM for α2A-, α2B-, and α2C-ARs, respectively); reverses decreases in systolic blood pressure induced by clonidine in spontaneously hypertensive rats at 10 mg/kg
Catalog No. 50-294-0642 Supplier Cayman Chemical Supplier No. 2762910 MG
An α2-AR antagonist (Kis = 5.4, 2, and 1.3 nM for α2A-, α2B-, and α2C-ARs, respectively); reverses decreases in systolic blood pressure induced by clonidine in spontaneously hypertensive rats at 10 mg/kg