Learn More
Cayman Chemical Piperoxan (hydrochloride), Purity: ≥98%, CAS: 135-87-5, 50 mg
SDP
Catalog No. 502940643
Encompass_Preferred
An α2-AR antagonist (Kis = 5.4, 2, and 1.3 nM for α2A-, α2B-, and α2C-ARs, respectively); reverses decreases in systolic blood pressure induced by clonidine in spontaneously hypertensive rats at 10 mg/kg
Catalog No. 50-294-0643 Supplier Cayman Chemical Supplier No. 2762950 MG
An α2-AR antagonist (Kis = 5.4, 2, and 1.3 nM for α2A-, α2B-, and α2C-ARs, respectively); reverses decreases in systolic blood pressure induced by clonidine in spontaneously hypertensive rats at 10 mg/kg