Learn More
Medchemexpress LLC PIPES (1,4-Piperazinediethanesulfonic acid) | 5625-37-6 | ≥99.0% | 5 G
SDP
Catalog No. 5000409870
Encompass_Preferred
PIPES (1,4-Piperazinediethanesulfonic acid) is an important component of PIPES buffer agent used in biochemistry. It is for research use only.
  • Important component of buffer agent
  • Prevents glutaraldehyde fixation-induced lipid loss and artifacts when used in buffer
Catalog No. 50-004-09870 Supplier Medchemexpress LLC Supplier No. HYD08755G
PIPES (1,4-Piperazinediethanesulfonic acid) is an important component of PIPES buffer agent used in biochemistry. It is for research use only.
  • Important component of buffer agent
  • Prevents glutaraldehyde fixation-induced lipid loss and artifacts when used in buffer