Learn More
Boston Bioproducts Inc PIPES Buffer (0.5 M, pH 6.8) - 100ml
Catalog No. NC9940565
Encompass
The PIPES Buffer is a zwitterionic buffering agent commonly used in biochemistry and molecular biology for its ability to maintain a stable pH. With a pKa of approximately 6.8, PIPES is effective in the pH range of 6.1 to 7.5, making it suitable for cell culture, enzyme assays, protein purification, and as a running buffer in gel electrophoresis. PIPES is also used in electron microscopy, protein crystallization, and isoelectric focusing and chromatography.
Catalog No. NC9940565 Supplier Boston Bioproducts Inc Supplier No. BB121100ML
Add to Cart
Add to Cart
The PIPES Buffer is a zwitterionic buffering agent commonly used in biochemistry and molecular biology for its ability to maintain a stable pH. With a pKa of approximately 6.8, PIPES is effective in the pH range of 6.1 to 7.5, making it suitable for cell culture, enzyme assays, protein purification, and as a running buffer in gel electrophoresis. PIPES is also used in electron microscopy, protein crystallization, and isoelectric focusing and chromatography.