Learn More
Boston Bioproducts Inc PIPES Buffer (0.5 M, pH 7.0) - 100ml
Catalog No. NC9083895
Encompass
The PIPES Buffer is a zwitterionic buffering agent commonly used in biochemistry and molecular biology for its ability to maintain a stable pH. With a pKa of approximately 6.8, PIPES is effective in the pH range of 6.1 to 7.5, making it suitable for cell culture, enzyme assays, protein purification, and as a running buffer in gel electrophoresis. PIPES is also used in electron microscopy, protein crystallization, and isoelectric focusing and chromatography.
Catalog No. NC9083895 Supplier Boston Bioproducts Inc Supplier No. BB122100ML
The PIPES Buffer is a zwitterionic buffering agent commonly used in biochemistry and molecular biology for its ability to maintain a stable pH. With a pKa of approximately 6.8, PIPES is effective in the pH range of 6.1 to 7.5, making it suitable for cell culture, enzyme assays, protein purification, and as a running buffer in gel electrophoresis. PIPES is also used in electron microscopy, protein crystallization, and isoelectric focusing and chromatography.