Learn More
Bioworld PIPES BUFFER 1M PH 6.0 100ML
Catalog No. NC2816889
Encompass_Preferred
liquid, 1.0m, ph 6Synonym(s): 1,4-Piperazinediethanesulfonic acidMolecular Formula: C8H18N2O6S2CAS Number: 5625-37-6Molecular Weight: 302.37
Catalog No. NC2816889 Supplier Bioworld Supplier No. 416200381
liquid, 1.0m, ph 6Synonym(s): 1,4-Piperazinediethanesulfonic acidMolecular Formula: C8H18N2O6S2CAS Number: 5625-37-6Molecular Weight: 302.37