Learn More
Medchemexpress LLC PIPES dipotassium | 108321-27-3 | MFCD00069751 | 98.0% | 378.55 | C8H16K2N2O6S2 | 10 G
SDP
Catalog No. 5000282178
Encompass_Preferred
PIPES dipotassium is a zwitterionic buffering agent used in biochemical and biological assays to maintain a stable, near-neutral pH. The dipotassium salt provides good aqueous solubility and is compatible with many enzymatic and cell-based applications.
  • Maintains pH in the 6.1-7.5 buffering range.
  • Zwitterionic structure minimizes metal binding and enzymatic interference.
  • High purity (98.0%).
  • White to off-white solid, water soluble.
  • Available in 10 G and larger lab-scale pack sizes.
  • Store at room temperature; keep under nitrogen and away from moisture; in solution store at -20°C to -80°C for long-term storage.
  • Supplied with SDS and COA documentation.
Catalog No. 50-002-82178 Supplier Medchemexpress LLC Supplier No. HYW32788610G
PIPES dipotassium is a zwitterionic buffering agent used in biochemical and biological assays to maintain a stable, near-neutral pH. The dipotassium salt provides good aqueous solubility and is compatible with many enzymatic and cell-based applications.
  • Maintains pH in the 6.1-7.5 buffering range.
  • Zwitterionic structure minimizes metal binding and enzymatic interference.
  • High purity (98.0%).
  • White to off-white solid, water soluble.
  • Available in 10 G and larger lab-scale pack sizes.
  • Store at room temperature; keep under nitrogen and away from moisture; in solution store at -20°C to -80°C for long-term storage.
  • Supplied with SDS and COA documentation.