Learn More
Medchemexpress LLC Pipes dipotassium | 108321-27-3 | MFCD00069751 | 98.0% | 378.55 | C8H16K2N2O6S2 | 100 G
SDP
Catalog No. 5000282059
Encompass_Preferred
PIPES (dipotassium) is the dipotassium salt of the zwitterionic buffering agent PIPES, provided as a white to off-white solid. It offers stable buffering near physiological pH with low metal ion binding, making it suitable for biochemical and cell biology applications such as assay buffers, electrophoresis, and sample preparation.
Catalog No. 50-002-82059 Supplier Medchemexpress LLC Supplier No. HYW327886100G
PIPES (dipotassium) is the dipotassium salt of the zwitterionic buffering agent PIPES, provided as a white to off-white solid. It offers stable buffering near physiological pH with low metal ion binding, making it suitable for biochemical and cell biology applications such as assay buffers, electrophoresis, and sample preparation.