Learn More
Medchemexpress LLC Pipes dipotassium | 108321-27-3 | MFCD00069751 | 98.0% | 378.55 g/mol | C8H16K2N2O6S2 | 50 G
SDP
Catalog No. 5000280665
Encompass_Preferred
PIPES (dipotassium) is the dipotassium salt of the zwitterionic buffering agent PIPES (1,4-piperazinediethanesulfonic acid). Supplied as a high-purity dry powder, it is used to prepare buffering solutions for biochemical and cell biology research where stable pH is required.
  • High purity: 98.0%.
  • Dipotassium salt form improves aqueous solubility.
  • Suitable for preparing pH-stable buffer solutions in biological assays.
  • Provided as a 50 g reagent pack for laboratory use.
  • Documentation available: data sheet, SDS, and COA.
Catalog No. 50-002-80665 Supplier Medchemexpress LLC Supplier No. HYW32788650G
Add to Cart
Add to Cart
PIPES (dipotassium) is the dipotassium salt of the zwitterionic buffering agent PIPES (1,4-piperazinediethanesulfonic acid). Supplied as a high-purity dry powder, it is used to prepare buffering solutions for biochemical and cell biology research where stable pH is required.
  • High purity: 98.0%.
  • Dipotassium salt form improves aqueous solubility.
  • Suitable for preparing pH-stable buffer solutions in biological assays.
  • Provided as a 50 g reagent pack for laboratory use.
  • Documentation available: data sheet, SDS, and COA.