Learn More
Research Products International Corp PIPES, Disodium Salt, 1 KG
SDP
Catalog No. 501977125
Encompass_Preferred
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5.
Catalog No. 50-197-7125 Supplier Research Products International Corp Supplier No. P401801000.0
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5.