Learn More
Research Products International Corp PIPES DISODIUM SALT, 100G
SDP
Catalog No. 50488923
Encompass_Preferred
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5.
Catalog No. 50-488-923 Supplier Research Products International Corp Supplier No. P40180100.0
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5.