Learn More
Research Products International Corp PIPES DISODIUM SALT, 500G
SDP
Catalog No. 50997959
Encompass_Preferred
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5.
Catalog No. 50-997-959 Supplier Research Products International Corp Supplier No. P40180500.0
Add to Cart
Add to Cart
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5.