Learn More
Research Products International Corp PIPES FREE ACID ULTRAPURE, 10GM
SDP
Catalog No. 50213620
Encompass_Preferred
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-213-620 Supplier Research Products International Corp Supplier No. P4014010.0
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5