Learn More
Research Products International Corp PIPES FREE ACID ULTRAPURE, 500GM
SDP
Catalog No. 50213624
Encompass_Preferred
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-213-624 Supplier Research Products International Corp Supplier No. P40140500.0
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5