Learn More
Research Products International Corp PIPES FREE ACID ULTRAPPURE, 50GM
SDP
Catalog No. 50213623
Encompass_Preferred
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-213-623 Supplier Research Products International Corp Supplier No. P4014050.0
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5