Learn More
Research Products International Corp PIPES FREE ACID ULTRAPURE, 1 KILOGRAM
SDP
Catalog No. 50488920
Encompass_Preferred
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-488-920 Supplier Research Products International Corp Supplier No. P401401000.0
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5