Learn More
Medchemexpress LLC Pipes (monosodium) | 10010-67-0 | 99.6% | 324.35 | 10 G
SDP
Catalog No. 5000451839
Encompass_Preferred
PIPES monosodium (1,4-Piperazinediethanesulfonic acid monosodium) is a pH buffer. It can be mixed with the disodium salt form of PIPES to adjust the pH and ionic strength of the medium by altering the ratio and total amounts of the buffers. This product is intended for research use only.
  • Functions as a pH buffer
  • Can be mixed with disodium salt form of PIPES
  • Allows adjustment of pH and ionic strength
  • Intended for research use only
Catalog No. 50-004-51839 Supplier Medchemexpress LLC Supplier No. HYW01127110G
PIPES monosodium (1,4-Piperazinediethanesulfonic acid monosodium) is a pH buffer. It can be mixed with the disodium salt form of PIPES to adjust the pH and ionic strength of the medium by altering the ratio and total amounts of the buffers. This product is intended for research use only.
  • Functions as a pH buffer
  • Can be mixed with disodium salt form of PIPES
  • Allows adjustment of pH and ionic strength
  • Intended for research use only