Learn More
Medchemexpress LLC PIPES (monosodium) | 10010-67-0 | 99.6% | 324.35 | 25 G
SDP
Catalog No. 5000366968
Encompass_Preferred
PIPES (1,4-Piperazinediethanesulfonic acid) monosodium is a pH buffer that can be mixed with another disodium salt form of PIPES. By changing the ratio and total amount of the PIPES monosodium and disodium buffers, the pH and ionic strength of the medium can be changed.
  • pH buffer
  • Can be mixed with disodium salt form of PIPES to adjust pH and ionic strength
  • For research use only
Catalog No. 50-003-66968 Supplier Medchemexpress LLC Supplier No. 25GHYW011271
PIPES (1,4-Piperazinediethanesulfonic acid) monosodium is a pH buffer that can be mixed with another disodium salt form of PIPES. By changing the ratio and total amount of the PIPES monosodium and disodium buffers, the pH and ionic strength of the medium can be changed.
  • pH buffer
  • Can be mixed with disodium salt form of PIPES to adjust pH and ionic strength
  • For research use only