Learn More
Research Products International Corp PIPES Sesquisdm Salt 250G
SDP
Catalog No. 502230493
Encompass_Preferred
PIPES Sesquisodium Salt, also known as PIPES Na3, is a chemical compound that has significant applications in the life science marketplace. PIPES stands for piperazine-N,N'-bis(2-ethanesulfonic acid), and the sesquisodium salt refers to the trisodium salt form of PIPES. 
It is primarily utilized as a buffering agent. Buffers are substances that help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for various enzymatic reactions and cellular activities. 
PIPES Sesquisodium Salt is particularly valuable in biological research, molecular biology, and cell culture applications. It offers excellent buffering capacity in the physiological pH range (6.1 - 7.5), making it suitable for a wide range of experimental conditions.
Catalog No. 50-223-0493 Supplier Research Products International Corp Supplier No. P52175250.0
PIPES Sesquisodium Salt, also known as PIPES Na3, is a chemical compound that has significant applications in the life science marketplace. PIPES stands for piperazine-N,N'-bis(2-ethanesulfonic acid), and the sesquisodium salt refers to the trisodium salt form of PIPES. 
It is primarily utilized as a buffering agent. Buffers are substances that help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for various enzymatic reactions and cellular activities. 
PIPES Sesquisodium Salt is particularly valuable in biological research, molecular biology, and cell culture applications. It offers excellent buffering capacity in the physiological pH range (6.1 - 7.5), making it suitable for a wide range of experimental conditions.