Learn More
Research Products International Corp PIPES Sodium Salt 500 G
SDP
Catalog No. 502230491
Encompass_Preferred
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-223-0491 Supplier Research Products International Corp Supplier No. P40150500.0
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5