Learn More
Research Products International Corp PIPES SODIUM SALT, 1 KILOGRAM
SDP
Catalog No. 50488922
Encompass_Preferred
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-488-922 Supplier Research Products International Corp Supplier No. P401501000.0
Add to Cart
Add to Cart
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5