Learn More
Research Products International Corp PIPES SODIUM SALT, 100 GRAM
SDP
Catalog No. 50488921
Encompass_Preferred
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Catalog No. 50-488-921 Supplier Research Products International Corp Supplier No. P40150100.0
pKa (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5