Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02003623 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-623 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-623 Supplier Avantor Supplier No. 426501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Specifications

CAS 5625-37-6
Quantity 100 g
Molecular Formula C8H18N2O6S2
Packaging Poly Bottle
Synonym pipes, 1,4-piperazinediethanesulfonic acid, piperazine-n,n'-bis 2-ethanesulfonic acid, 2,2'-piperazine-1,4-diyl diethanesulfonic acid, 1,4-piperazinebis ethanesulfonic acid, unii-g502h79v6l, 2-4-2-sulfoethyl piperazin-1-yl ethanesulfonic acid, piperazine-1,4-bis 2-ethanesulfonic acid, 1,4-piperazine-diethanesulfonic acid, 2,2'-piperazine-1,4-diyl bis ethanesulphonic acid
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
Molecular Weight (g/mol) 302.36
PubChem CID 79723
ChEBI CHEBI:44933
Formula Weight 302.37
Grade Ultrapure Bioreagent
Chemical Name or Material PIPES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.