Learn More
Hach Company Pipette, Variable Volume, 0.2 - 1.0 mL
Supplier:  Hach Company BBP078
Encompass
Hach's variable volume single-channel, air-displacement pipettes feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation. Outstanding accuracy with adjustments in 50 μL increments (0.05 mL). Ideal for use with Hach TNTplus reagents. Supplied with calibration report and maintenance tool.Pipette tips sold separately.
Catalog No. 50-199-7546