Learn More
Quality Biological Inc Serological Pipet, 1 mL, Polystryrene, Individually wrapped, Sterile, packed 100/bag, 800/CS,
SDP
Catalog No. 507517907
Encompass_Preferred
Serological Pipet, 1 mL, Polystyrene, Individually Wrapped in Paper/Plastic, Packed in Bags. Sterile (gamma irradiated).
Packaged 100/Bag, 800/CS.
• Manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Sterile: Yes
Maximum Volume: 1 mL
Graduated: Yes
Graduation Increments (Subdivisions): 1/100 mL
Material: Polystyrene
Catalog No. 50-751-7907 Supplier Quality Biological Inc Supplier No. P886229201B
Serological Pipet, 1 mL, Polystyrene, Individually Wrapped in Paper/Plastic, Packed in Bags. Sterile (gamma irradiated).
Packaged 100/Bag, 800/CS.
• Manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Sterile: Yes
Maximum Volume: 1 mL
Graduated: Yes
Graduation Increments (Subdivisions): 1/100 mL
Material: Polystyrene