Learn More
Chemglass Life Sciences Pipet, 1mL, Volumetric, To Deliver and To Contain Markings
SDP
Supplier:  Chemglass Life Sciences CG87081
Encompass_Preferred
Pipet, 1mL, Volumetric, To Deliver and To Contain Markings. These precision pipets are manufactured and calibrated “To Contain” (bottom line) and “To Deliver” (top line) to specifications for Class A volumetric ware. Do not pipet by mouth. We suggest using a mechanical pipetting device.
Catalog No. NC1433059