Learn More
Chemglass Life Sciences Pipet, 20mL, Volumetric, To Deliver and To Contain Markings
SDP
Catalog No. NC1433063
Encompass_Preferred
Pipet, 20mL, Volumetric, To Deliver and To Contain Markings. These precision pipets are manufactured and calibrated “To Contain” (bottom line) and “To Deliver” (top line) to specifications for Class A volumetric ware. Do not pipet by mouth. We suggest using a mechanical pipetting device.
Catalog No. NC1433063 Supplier Chemglass Life Sciences Supplier No. CG870820
Pipet, 20mL, Volumetric, To Deliver and To Contain Markings. These precision pipets are manufactured and calibrated “To Contain” (bottom line) and “To Deliver” (top line) to specifications for Class A volumetric ware. Do not pipet by mouth. We suggest using a mechanical pipetting device.