Learn More
Chemglass Life Sciences 25.0mL Pipette Pump, Minum-Ware
SDP
Catalog No. 50872312
Encompass_Preferred
25.0mL Pipette Pump, Minum-Ware. Provides for safe, convenient and trouble free pipetting. Rotation of the thumbwheel fills or dispenses the volume required from the pipette. The top plunger can be depressed for rapid emptying. A flexible, tapered chuck allows the pipette to fit smoothly and securely into the pump. NOTE: Part Number MW-126-02 fits both sizes of the MW-124 Pasteur pipettes.
Catalog No. 50-872-312 Supplier Chemglass Life Sciences Supplier No. MW12604
25.0mL Pipette Pump, Minum-Ware. Provides for safe, convenient and trouble free pipetting. Rotation of the thumbwheel fills or dispenses the volume required from the pipette. The top plunger can be depressed for rapid emptying. A flexible, tapered chuck allows the pipette to fit smoothly and securely into the pump. NOTE: Part Number MW-126-02 fits both sizes of the MW-124 Pasteur pipettes.