Learn More
Quality Biological Inc Serological Pipet, 25 mL, Polystyrene, Individually wrapped, Sterile, packed 50/bag, 200/CS
SDP
Catalog No. 507517909
Encompass_Preferred
Serological Pipet, 25 mL, Polystyrene, Individually wrapped, Sterile
Packed 50 Individually Wrapped/bag, 200/CS
• All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification
Catalog No. 50-751-7909 Supplier Quality Biological Inc Supplier No. P886229225B
Serological Pipet, 25 mL, Polystyrene, Individually wrapped, Sterile
Packed 50 Individually Wrapped/bag, 200/CS
• All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934
• Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink
• Premium material barrier plug
• Color coded stripes and packaging enable quick volume identification